Anti-Histiocytoma Marker Mouse Monoclonal Antibody (CF594) [clone: D11]
Supplier: Biotium
BNC940348-500 BNC940348-100
BNC940348-500EA 560 EUR
BNC940348-500 BNC940348-100
Anti-Histiocytoma Marker Mouse Monoclonal Antibody (CF594) [clone: D11]
Antibodies
In Western blotting, this antibody detects an antigen of 125 kDa in human liver and 135 kDa in tumors of histiocytic origin. Comparative study of this MAb and a standard CD68 MAb showed that their antigens are different. Its antigen in all macrophage types studied is located on the plasma membrane and within cytoplasmic structures including lysosomes. This MAb shows a restricted reactivity to cells of the monocyte/macrophage system. It specifically reacts with blood monocytes and stains resident macrophages in a wide variety of human tissues. This MAb does not stain antigen-presenting cells,More Product Information