Anti-Bovine Serum Albumin Mouse Monoclonal Antibody (Biotin) [clone: ALB/398]

Supplier: Biotium
BNCB0398-500 BNCB0398-100
BNCB0398-500EA 560 EUR
BNCB0398-500 BNCB0398-100
Anti-Bovine Serum Albumin Mouse Monoclonal Antibody (Biotin) [clone: ALB/398]
Antibodies
Reacts with a protein of ~66 kDa, identified as bovine serum albumin (BSA). It is a high affinity antibody and can be used for detection of traces of BSA. Bovine serum albumin (BSA) is an abundant plasma protein in cows that is important for maintaining osmotic pressure in blood plasma for proper distribution of body fluids between intravascular compartments and body tissues. BSA is a common buffer component for immunoglobulin type assays due to good solubility characteristics for water, Ca2 , Na , K , fatty acids, hormones and bilirubin. BSA makes up about half of the protein in plasma and represents the most stable and soluble protein in the plasma. It is a suitable reagent for laboratories developing immunoassays, mostly due to its availability, solubility and the numerous functional groups present for coupling. The BSA component contains several lysines that are capable of reacting with conjugation sites of linkers, making it applicable as a carrier protein for antigenic compounds.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: ALB/398
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR