Anti-UPK3A Rabbit Polyclonal Antibody (CF405S) [clone: Rabbit PAb]

Supplier: Biotium
BNC040409-500 BNC040409-100
BNC040409-500EA 560 EUR
BNC040409-500 BNC040409-100
Anti-UPK3A Rabbit Polyclonal Antibody (CF405S) [clone: Rabbit PAb]
Antibodies
Uroplakin-3A (UPK3A) is a component of the asymmetric unit membrane (AUM). It is a highly specialized bio-membrane made by terminally differentiated urothelial cells. The protein may play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. UPK3A also contributes to the formation of urothelial glycocalyx, which may play an important role in preventing bacterial adherence through FimH bacterial protein binding leading to bladder infection. UPK3A has been shown as a helpful marker for the detection of bladder cancer.

Type: Primary
Antigen: UPK3A
Clonality: Polyclonal
Clone: Rabbit PAb
Conjugation: CF405S
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR