Anti-SLC3A2 Mouse Monoclonal Antibody (CF405S) [clone: UM7F8]

Supplier: Biotium
BNC040425-500 BNC040425-100
BNC040425-500EA 560 EUR
BNC040425-500 BNC040425-100
Anti-SLC3A2 Mouse Monoclonal Antibody (CF405S) [clone: UM7F8]
Antibodies
CD98 exits as a heterodimer containing a disulphide-linked glycosylated heavy chain and a non-glycosylated light chain. It is a member of the solute carrier family and encodes a cell surface, transmembrane protein. The protein exists as the heavy chain of a heterodimer, covalently bound through disulfide bonds to one of several possible light chains. The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids. Alternatively spliced transcript variants, encoding different isoforms, have been characterized.

Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: UM7F8
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR