Anti-CD44 Mouse Monoclonal Antibody (CF405S) [clone: 156-3C11]

Supplier: Biotium
BNC040460-500 BNC040460-100
BNC040460-500EA 560 EUR
BNC040460-500 BNC040460-100
Anti-CD44 Mouse Monoclonal Antibody (CF405S) [clone: 156-3C11]
Antibodies
Recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. This MAb selectively interferes with lymphocyte binding to lymph node, mucosal and synovial endothelium. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD44 CD44
Antigen name CD44 Standard CD44 Standard
Environmentally Preferable
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation CF® 405S CF® 405S
Clone Clone 156-3C11 Clone 156-3C11
Reactivity Baboon,Green monkey,Human Baboon,Green monkey,Human
Host Mouse Mouse
Gene ID 502328 502328
Isotype IgG2a kappa IgG2a kappa
Western blot Yes Yes
ImmunoChemistry Yes Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR