Anti-PGR Mouse Monoclonal Antibody (CF594) [clone: PR501]

Supplier: Biotium
BNC940501-500 BNC940501-100
BNC940501-500EA 560 EUR
BNC940501-500 BNC940501-100
Anti-PGR Mouse Monoclonal Antibody (CF594) [clone: PR501]
Antibodies
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR501
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol PGR
Antigen name Progesterone Receptor
Environmentally Preferable
Antigen synonyms PR|PRA|Progesterone receptor form A|Nuclear receptor subfamily 3 group C member 3|NR3C3|PRB|PGR|Progesterone receptor form B
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 594
Clone Clone PR501
Reactivity Human
Host Mouse
Gene ID 2905
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR