Anti-VWF Mouse Monoclonal Antibody (Biotin) [clone: 3E2D10]

Supplier: Biotium
BNCB0633-500 BNCB0633-100
BNCB0633-500EA 560 EUR
BNCB0633-500 BNCB0633-100
Anti-VWF Mouse Monoclonal Antibody (Biotin) [clone: 3E2D10]
Antibodies
von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascularMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol VWF
Environmentally Preferable
Antigen name von Willebrand Factor
Antigen synonyms F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2
Antibody type Primary
Clonality Monoclonal
Conjugation Biotin
Clone Clone 3E2D10
Reactivity Human
Host Mouse
Gene ID 440848
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR