Anti-VWF Mouse Monoclonal Antibody (CF640R) [clone: VWF635]

Supplier: Biotium
BNC400635-500 BNC400635-100
BNC400635-500EA 560 EUR
BNC400635-500 BNC400635-100
Anti-VWF Mouse Monoclonal Antibody (CF640R) [clone: VWF635]
Antibodies
von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascular lesions from those of other tissue differentiation within a panel of other vascular markers although not all tumors of endothelial differentiation contain this antigen.

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: VWF635
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR