Anti-NKX2-1 Mouse Monoclonal Antibody (CF647) [clone: NX2.1/690]

Supplier: Biotium
BNC470690-500 BNC470690-100
BNC470690-500EA 560 EUR
BNC470690-500 BNC470690-100
Anti-NKX2-1 Mouse Monoclonal Antibody (CF647) [clone: NX2.1/690]
Antibodies
Recognizes a protein of 40 kDa, identified as Thyroid transcription factor-1 (TTF-1). TTF-1 is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. Anti-TTF-1 is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the breast, mediastinal germ cell tumors, and malignant mesothelioma. It can also be used to differentiate small cell lung carcinoma from lymphoid infiltrates.Loss of TTF-1 expression in non-small cell lung carcinoma has been associated with aggressive behavior of such neoplasms. TTF-1 reactivity is also seen in thyroid malignancies.

Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: NX2.1/690
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR