Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF568) [clone: MTC719]
Supplier: Biotium
BNC680719-500 BNC680719-100
BNC680719-500EA 560 EUR
BNC680719-500 BNC680719-100
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF568) [clone: MTC719]
Antibodies
This MAb recognizes a 60 kDa antigen associated with the mitochondria in cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in cells from a wide variety of animals, but not insects and bacteria. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-More Product Information