Anti-KRT19 Mouse Monoclonal Antibody (CF405S) [clone: A53-B/A2.26 BA17]
Supplier: Biotium
BNC040753-500 BNC040753-100
BNC040753-500EA 560 EUR
BNC040753-500 BNC040753-100
Anti-KRT19 Mouse Monoclonal Antibody (CF405S) [clone: A53-B/A2.26 BA17]
Antibodies
This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40 kDa. CK19 is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled.More Product Information