Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF594) [clone: MTC754]

Supplier: Biotium
BNC940754-500 BNC940754-100
BNC940754-500EA 560 EUR
BNC940754-500 BNC940754-100
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF594) [clone: MTC754]
Antibodies
This MAb recognizes a 60 kDa antigen associated with the mitochondria in cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in cells from a wide variety of animals including insects and bacteria. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells and may be used to stain mitochondria of cells in frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.

Type: Primary
Antigen: Mitochondrial Marker
Clonality: Monoclonal
Clone: MTC754
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR