Anti-CNN1 Mouse Monoclonal Antibody [clone: CALP CNN1/832]
Supplier: Biotium
BNUB0833-500 BNUB0833-100
BNUB0833-500EA 511 EUR
BNUB0833-500 BNUB0833-100
Anti-CNN1 Mouse Monoclonal Antibody [clone: CALP CNN1/832]
Antibodies
Multiple isoelectric variants of calponin have been identified, however only two molecular weight isoforms exist; a 34 kDa form and a 29 kDa form. Expression of the 29 kDa form, I-calponin, is primarily restricted to muscle of the urogenital tract, whereas the higher molecular weight variant has been demonstrated in vascular and visceral smooth muscle. In Western blotting, this MAb reacts with only the 34 kDa form of calponin in extracts of human aortic medial smooth muscle and is unreactive with fibroblast extracts of cultivated human foreskin. Calponin is a calmodulin, F-actin andMore Product Information