Anti-RBP1 Mouse Monoclonal Antibody (CF405S) [clone: RBP/872]
Supplier: Biotium
BNC040872-500 BNC040872-100
BNC040872-500EA 560 EUR
BNC040872-500 BNC040872-100
Anti-RBP1 Mouse Monoclonal Antibody (CF405S) [clone: RBP/872]
Antibodies
Recognizes a protein of 21 kDa-25 kDa, identified as retinol binding protein-1 (RBP1). This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post-transnationally and results in defective delivery and supply to the epidermal cells.
Type: Primary
Antigen: RBP1
More Product Information