Anti-AMPD3 Mouse Monoclonal Antibody (Biotin) [clone: AMPD3/901]
Supplier: Biotium
BNCB0901-500 BNCB0901-100
BNCB0901-500EA 560 EUR
BNCB0901-500 BNCB0901-100
Anti-AMPD3 Mouse Monoclonal Antibody (Biotin) [clone: AMPD3/901]
Antibodies
This antibody recognizes a protein of ~90 kDa, which is identified as Adenosine Monophosphate Deaminase, isoform E (AMPD3). It has 767 amino acids and is assigned an EC 3.5.4.6. It is a highly regulated enzyme that catalyzes the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. AMPD3 gene encodes the erythrocyte (E) isoforms, whereas other family members encode isoforms that predominate in muscle (M) and liver (L) cells. This MAb shows reactivity with cells of the erythroid lineage at all stages of maturation in theMore Product Information