Anti-NCL Mouse Monoclonal Antibody (CF488A) [clone: NCL/902]

Supplier: Biotium
BNC880902-500 BNC880902-100
BNC880902-500EA 560 EUR
BNC880902-500 BNC880902-100
Anti-NCL Mouse Monoclonal Antibody (CF488A) [clone: NCL/902]
Antibodies
Recognizes a protein of ~76 kDa, which is identified as Nucleolin (NCL). It is the major nucleolar phosphoprotein of growing eukaryotic cells. NCL is located mainly in dense fibrillar regions of the nucleolus. It is found associated with intranucleolar chromatin and pre-ribosomal particles. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly.This MAb can be used to stain the nucleoli in cell or tissue preparations and can be used as a marker of the nucleoli in subcellular fractions. It produces a speckled pattern in the nuclei of cells of normal and malignant cells and may be used to stain the nucleoli of cells in fixed or frozen tissue sections. It can be used with paraformaldehyde fixed frozen tissue or cell preparations and formalin fixed, paraffin-embedded tissue sections.

Type: Primary
Antigen: NCL
Clonality: Monoclonal
Clone: NCL/902
Conjugation: CF488A
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG1, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR