Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF640R) [clone: AE-1]

Supplier: Biotium
BNC400905-500 BNC400905-100
BNC400905-500EA 560 EUR
BNC400905-500 BNC400905-100
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF640R) [clone: AE-1]
Antibodies
This MAb recognizes a 60 kDa antigen associated with the mitochondria in human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in human cells only. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignantMore Product Information
Order Now

SPECIFICATIONS

Antigen name Mitochondrial Marker
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 640R
Clone Clone AE-1
Reactivity Human
Host Mouse
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR