Anti-CD44 Mouse Monoclonal Antibody (CF405S) [clone: HCAM/918]

Supplier: Biotium
BNC040918-500 BNC040918-100
BNC040918-500EA 560 EUR
BNC040918-500 BNC040918-100
Anti-CD44 Mouse Monoclonal Antibody (CF405S) [clone: HCAM/918]
Antibodies
Recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cellMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD44
Antigen name CD44 Standard
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone Clone HCAM/918
Reactivity Primates,Human
Host Mouse
Gene ID 502328
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR