Anti-CD46 Mouse Monoclonal Antibody (CF640R) [clone: 197-2B1]

Supplier: Biotium
BNC400931-500 BNC400931-100
BNC400931-500EA 560 EUR
BNC400931-500 BNC400931-100
Anti-CD46 Mouse Monoclonal Antibody (CF640R) [clone: 197-2B1]
Antibodies
Recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virus receptor, an adherence factor for group A Streptococcus pyogenes, and a cellular pilus receptor for pathogenic Neisseria. This MAb can be applied to test complement activation in pseudo-allergic reactions to acetylsalicylic acid and to test for measles virus infection of cells.

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: 197-2B1
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR