Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: MUC1/967]

Supplier: Biotium
BNC940967-500 BNC940967-100
BNC940967-500EA 560 EUR
BNC940967-500 BNC940967-100
Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: MUC1/967]
Antibodies
This MAb recognizes full-length MUC1 in a glycosylation-independent manner and can bind to the fully glycosylated protein. The dominant epitope of this MAb is APDTR in the VNTR region. It reacts with the core peptide of the MUC1 protein, which is a member of a family of mucin glycoproteins that are characterized by high carbohydrate content, O-linked oligosaccharides, high molecular weight (>200 kDa) and an amino acid composition rich in serine, threonine, proline and glycine. The core protein contains a domain of 20 amino-acid tandem repeats that functions as multiple epitopes for theMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol MUC1
Environmentally Preferable
Antigen name Mucin 1 / EMA / Episialin / CD227
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 594
Clone Clone MUC1/967
Reactivity Human
Host Mouse
Gene ID 89603
Isotype IgG1
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR