Anti-TFGB Mouse Monoclonal Antibody (CF770) [clone: 1D11.16.8]

Supplier: Biotium
BNC700977-500 BNC700977-100
BNC700977-500EA 560 EUR
BNC700977-500 BNC700977-100
Anti-TFGB Mouse Monoclonal Antibody (CF770) [clone: 1D11.16.8]
Antibodies
This MAb recognizes TGF beta 1, 2 and 3. Three TGF betas have been identified in mammals. TGF beta-1, TGF beta-2 and TGF beta-3 are each synthesized as precursor proteins that are very similar in that each is cleaved to yield a 112 amino acid polypeptide that remains associated with the latent portion of the molecules. Biologically active TGF betarequires dimerization of the monomers (usually homodimers) and release of the latent peptide portion. Overall, the mature region of the TGF beta-3 protein has approximately 80% identity to the mature region of both TGF beta-1 and TGF beta-2. However, the NH2 terminals or precursor regions of their molecules share only 27% sequence identity. TGF betas inhibit the growth of epithelial cells and stimulate the growth of mesenchymal cells.

Type: Primary
Antigen: TFGB
Clonality: Monoclonal
Clone: 1D11.16.8
Conjugation: CF770
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Dog, Hamster, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR