Anti-ITGA2B Mouse Monoclonal Antibody (CF594) [clone: ITGA2B/1036]

Supplier: Biotium
BNC941036-500 BNC941036-100
BNC941036-500EA 560 EUR
BNC941036-500 BNC941036-100
Anti-ITGA2B Mouse Monoclonal Antibody (CF594) [clone: ITGA2B/1036]
Antibodies
Reacts with a calcium-dependent complex of CD41/CD61, a dimer of 90 kDa and 140 kDa present on the membrane of normal platelets and megakaryocytes. CD41/CD61 is also known as platelet glycoprotein GPIIb/GPIIIa or integrin IIa/3. This complex is the receptor of fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation.

Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: ITGA2B/1036
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ITGA2B
Antigen name CD41a
Environmentally Preferable
Antigen synonyms Integrin alpha IIb|platelet fibrinogen receptor|alpha subunit|HPA3|BDPLT2|platelet glycoprotein IIb of IIb/IIIa complex|ITGAB|BDPLT16|GT|ITGA2B|GPalpha IIb
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 594
Clone Clone ITGA2B/1036
Reactivity Human
Host Mouse
Gene ID 411312
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR