Anti-ITGB3 Mouse Monoclonal Antibody (Purified) [clone: 2.50E 12]

Supplier: Biotium
BNUB1048-500 BNUB1048-100
BNUB1048-500EA 511 EUR
BNUB1048-500 BNUB1048-100
Anti-ITGB3 Mouse Monoclonal Antibody (Purified) [clone: 2.50E 12]
Antibodies
Reacts with human integrin beta3 (GPIIIa, vitronectin receptor beta chain). The protein detectable is a complex of CD41 and CD61.The apparent molecular weight of the GPIIIa by SDS-PAGE is 105 kDa reduced and 90 kDa unreduced. Ligands are fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin. Residues 237-248 of GPIIIa or CD61 are critical in adhesive protein binding.

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: 2.50E 12
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR