Anti-CD3E Mouse Monoclonal Antibody (CF488A) [clone: B-B12]

Supplier: Biotium
BNC881052-500 BNC881052-100
BNC881052-500EA 560 EUR
BNC881052-500 BNC881052-100
Anti-CD3E Mouse Monoclonal Antibody (CF488A) [clone: B-B12]
Antibodies
Reacts with five invariable CD3 chains (designated as and ) with molecular weight ranging from 16-28 kDa. CD3 is expressed, typically at high levels, on peripheral T cells and majority of T cell neoplasms. Thymocytes express CD3 at different level on the cell surface in the course of differentiation and, in cortical thymus, CD3 is predominantly Intracytoplasmic. The CD3 complex is closely associated at the lymphocyte cell surface with T cell antigen receptor (TCR) and is involved in transducing antigen-recognition signals into cytoplasm of T cells and in regulating the cell surfaceMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD3E
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone Clone B-B12
Reactivity Human
Host Mouse
Gene ID 3003
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR