Anti-ABO Mouse Monoclonal Antibody (Purified) [clone: HE-10]

Supplier: Biotium
BNUB1089-500 BNUB1089-100
BNUB1089-500EA 511 EUR
BNUB1089-500 BNUB1089-100
Anti-ABO Mouse Monoclonal Antibody (Purified) [clone: HE-10]
Antibodies
This MAb preferably reacts with determinants of chain A and H type 3(Gal1-3GalNAc-R) and 4 (Gal1-3GalNAc-R), but not with type 1 and 2 chain structures. It is not reactive with immuno-dominant A trisaccharide. This MAb is applicable for tissue staining in tumor patients with blood groups A and AB. It shows a highly heterogeneous reactivity in human colon tumor tissue and adjacent mucosa. Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelialMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ABO
Environmentally Preferable
Antigen name Blood Group A
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Clone HE-10
Reactivity Human
Host Mouse
Isotype IgM kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,2 mg/ml
Storage buffer PBS/0,05% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR