HA-100 hydrochloride ≥95%

Supplier: Cayman Chemical

14955-25 14955-5 14955-10 14955-50
CAYM14955-25EA 401 EUR
CAYM14955-25 CAYM14955-5 CAYM14955-10 CAYM14955-50
HA-100 hydrochloride ≥95%
HA-100 hydrochloride

HA-100 is an isoquinoline compound with an added piperazinylsulfonyl group that acts as an inhibitor of protein kinases (PKs), including PKA, PKC, and PKG (IC50s = 8, 12 and 4 µM, respectively). It less effectively blocks the activity of myosin light chain kinase (IC50 = 240 µM).

MW: 350.3 g/mol
Storage Temperature: Freezer
CAS Number: 210297-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR