Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, white ≥97.0%, solid

Supplier: Thermo Scientific

Synonyms: TBTA, Tris((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)amine

H66485.03 H66485.MD
H66485.03EA 500 EUR
H66485.03 H66485.MD
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, white ≥97.0%, solid
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilises Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.


  • Incompatible with heat and oxidising agents
  • Store at −20 °C

Formula: C₃₀H₃₀N₁₀
MW: 530,64 g/mol
Melting Pt: 132…143 °C
Storage Temperature: Freezer
MDL Number: MFCD09265124
CAS Number: 510758-28-8

Order Now

Specification Test Results

Appearance (Color) White
Form Solid
Assay by NMR 97,0%
Solubility Soluble in DMSO, DMF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR