Autoclaves, Touchclave®-Lab 'K' series
Supplier: LTE SCIENTIFIC
TC/150/KE1 TC/150/KS6 TC/200/KE1 TC/200/KS6 TC/300/KE1 TC/300/KS6 TC/360/KE1 TC/360/KS6 TC/450/KE1 TC/450/KS6
481-0295EA 56810 EUR
481-0295 481-0297 481-0333 481-0334 481-0336 481-0337 481-0650 481-0652 481-0651 481-0653
Autoclaves, Touchclave®-Lab 'K' series
Sterilisers Autoclaves (Steam Sterilisers)
‘K’ series models are designed to operate using either a steam generator situated beneath the chamber, or via the customer's own direct steam supply. This arrangement maximises the usable chamber volume and provides a clean, unencumbered chamber, free from heaters and water.
  • Eight program touch screen control system
  • Internal data archiving for up to 5000 cycles (depending on configuration)
  • Water jacketed chamber for exceptional cooling efficiency
  • Water conservation system and exhaust condensate
  • Integral silent air compressor
All models incorporate anMore Product Information