Coal fly ash - constituents
Supplier: LGC Standards PROMOCHEM
NIST-2689
NIST-2690
NIST-2691
PROONIST-2689EA
1030
EUR
PROONIST-2689
PROONIST-2690
PROONIST-2691
Coal fly ash - constituents
Standards
Petroleum Industry Standards
These reference standard is intended for use in the evaluation of analytical methods used for the classification of coal fly ash and for the determination of constituent elements in coal fly ash or materials of a similar matrix.
A unit consists of three 10 g hermetically sealed glass vials of fly ash pulverised to less than 145 µm particle size and blended to a high degree of homogeneity.
Certifications: Traceable to NIST and SRM.