2-Aminopyridine-6-methanol 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(6-Aminopyridin-2-yl)methanol
450040010
450040050
ACRO450040010EA
195
EUR
ACRO450040010
ACRO450040050
2-Aminopyridine-6-methanol 97%
2-Aminopyridine-6-methanol
Formula:
C₆H₈N₂O MW: 124,14 g/mol Melting Pt: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD09863097 CAS Number: 79651-64-2 |
Specification Test Results
Appearance | Off-white to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 89 to 95 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...