Amodiaquine dihydrochloride dihydrate

Supplier: Thermo Scientific
Warning

Synonyms: 4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate, 4-[(7-Chloroquinolin-4-yl)amino]-2-[(diethylamino)methyl]phenol dihydrochloride

455200050 455200250
ACRO455200050EA 48.8 EUR
ACRO455200050 ACRO455200250
Amodiaquine dihydrochloride dihydrate
Amodiaquine dihydrochloride dihydrate
Formula: C20H22ClN3O .2HCl .2H2O
MW: 464,812 g/mol (392,33 anhy)
Boiling Pt: 478 °C (760 mmHg)
Melting Pt: 166…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00078857
CAS Number: 6398-98-7

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 169 °C
Water 7.0 to 9.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR