Topiramate 98%

Supplier: Thermo Scientific
Warning

Synonyms: 2,3:4,5-Di-O-isopropylidene-1-O-sulphamoyl-beta-D-fructopyranose, ((3aS,5aR,8aR,8bS)-2,2,7,7-Tetramethyltetrahydro-3aH-bis([1,3]dioxolo)[4,5-b:4',5'-d]pyran-3a-yl)methyl sulphamate

456190010 456190050
ACRO456190010EA 98.2 EUR
ACRO456190010 ACRO456190050
Topiramate 98%
Topiramate
Formula: C₁₂H₂₁NO₈S
MW: 339,36 g/mol
Melting Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00865320
CAS Number: 97240-79-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 124 to 128 °C
HPLC ≥97.5 % (ELSD)
Specific optical rotation -29° to -33° (20 °C, 589 nm) (c=1,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR