Risperidone 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-(2-(4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl)ethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one

459390010 459390050
ACRO459390010EA 59.2 EUR
ACRO459390010 ACRO459390050
Risperidone 99%
Risperidone

Soluble in chloroform, methanol.

Formula: C₂₃H₂₇FN₄O₂
MW: 410.49 g/mol
Melting Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00274576
CAS Number: 106266-06-2
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 170 to 174 °C
Loss on drying ≤0.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR