Octylamine 99+%

Supplier: Thermo Fisher Scientific
Danger

129490050 129495000 129491000 129490020
ACRO129490050EA 41.5 EUR
ACRO129490050 ACRO129495000 ACRO129491000 ACRO129490020
Octylamine 99+%
Octylamine
Formula: H₃C(CH₂)₆CH₂NH₂
Boiling Pt: 179 °C (760 mmHg)
Melting Pt: –1 °C
Density: 0,782 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008247
CAS Number: 111-86-4
UN: 2734
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.0 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR