Miconazole nitrate
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate
291830010
291830050
291830250
ACRO291830010EA
54.3
EUR
ACRO291830010
ACRO291830050
ACRO291830250
Miconazole nitrate
Miconazole nitrate
Formula:
C₁₈H₁₄Cl₄N₂O·HNO₃ MW: 479,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00058161 CAS Number: 22832-87-7 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C, 2 h) |
HPLC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...