Calcium iodate, pure
Supplier: Thermo Scientific
|
Danger
|
318410025
318411000
318415000
ACRO318410025EA
408
EUR
ACRO318410025
ACRO318411000
ACRO318415000
Calcium iodate, pure
Calcium iodate
Formula:
CaI₂O₆ MW: 389,881 g/mol Melting Pt: 540 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00036265 CAS Number: 7789-80-2 UN: 1479 ADR: 5.1,II |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Titration Iodometric | ≥96 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...