5-Amino-2-(methylthio)pyrimidine 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-(Methylthio)-5-pyrimidinamine
449820010
449822500
ACRO449820010EA
370
EUR
ACRO449820010
ACRO449822500
5-Amino-2-(methylthio)pyrimidine 96%
5-Amino-2-(methylthio)pyrimidine
Formula:
C₅H₇N₃S Storage Temperature: Ambient |
MDL Number:
MFCD10697095 CAS Number: 42382-46-7 |
Specification Test Results
Appearance | Orange-red to red Powder |
Infrared spectrum | Conforms |
HPLC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...