1-Butylimidazole 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Butyl-1H-imidazole, 1-Butyl-1H-imidazole, N-Butylimidazole

442770250 442771000
ACRO442770250EA 69 EUR
ACRO442770250 ACRO442771000
1-Butylimidazole 98%
1-Butylimidazole
Formula: C₇H₁₂N₂
Boiling Pt: 257 °C (760 mmHg)
Density: 0,945 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042753
CAS Number: 4316-42-1
UN: 2690
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4810 (20°C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR