2-Chlorothiazole 97%
Supplier: Thermo Scientific
|
Warning
|
457260010
457260050
457260250
ACRO457260010EA
40
EUR
ACRO457260010
ACRO457260050
ACRO457260250
2-Chlorothiazole 97%
2-Chlorothiazole
Formula:
C₃H₂ClNS MW: 119,57 g/mol Boiling Pt: 145…146 °C (760 mmHg) Density: 1,378 g/cm³ Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210701 CAS Number: 3034-52-4 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to yellow to light brown Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...