Gemcitabine 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

456890010 456890050
ACRO456890010EA 123 EUR
ACRO456890010 ACRO456890050
Gemcitabine 98%
Gemcitabine
Formula: C₉H₁₁F₂N₃O₄
MW: 263.2 g/mol
Melting Pt: 292 °C
Storage Temperature: Freezer
MDL Number: MFCD00869720
CAS Number: 95058-81-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR