Coenzyme Q10 98%

Supplier: Thermo Fisher Scientific

457950010 457950050 457950250
ACRO457950010EA 65.5 EUR
ACRO457950010 ACRO457950050 ACRO457950250
Coenzyme Q10 98%
Coenzyme Q10
Formula: C₅₉H₉₀O₄
Melting Pt: 49 °C
Storage Temperature: Freezer
MDL Number: MFCD00042919
CAS Number: 303-98-0

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific absorbance A (1%/1cm) ≥162 (275 nm, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR