Melphalan 94%

Supplier: Thermo Fisher Scientific
Danger

458650010 458650050
ACRO458650010EA 219 EUR
ACRO458650010 ACRO458650050
Melphalan 94%
Melphalan
Formula: C₁₃H₁₈Cl₂N₂O₂
Melting Pt: 180 °C
Storage Temperature: Freezer
CAS Number: 148-82-3
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥93 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR