7-Hydroxy-1,2,3,4-tetrahydroquinoline 96%

Supplier: Thermo Scientific
Warning

458710010 458710050
ACRO458710010EA 90.2 EUR
ACRO458710010 ACRO458710050
7-Hydroxy-1,2,3,4-tetrahydroquinoline 96%
7-Hydroxy-1,2,3,4-tetrahydroquinoline
Formula: C₉H₁₁NO
MW: 149,1897 g/mol
Melting Pt: 91…93,5 °C
Storage Temperature: Ambient
MDL Number: MFCD08063887
CAS Number: 58196-33-1

Order Now

Specification Test Results

Appearance White to light brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 89 to 95 °C
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR