4-Bromoisoxazole 95%

Supplier: Thermo Scientific
Warning

458730010 458730050
ACRO458730010EA 36.7 EUR
ACRO458730010 ACRO458730050
4-Bromoisoxazole 95%
4-Bromoisoxazole
Formula: C₃H₂BrNO
MW: 147,9600 g/mol
Melting Pt: 39…41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179569
CAS Number: 97925-43-4

Order Now

Specification Test Results

Appearance White to yellow Low melting solid
Infrared spectrum Conforms
Melting point 37 to 43 °C
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR