Acarbose 95%
Supplier: Thermo Scientific
Synonyms:
Prandase, Acarbose Hydrate, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose, Precose, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4S,5S,6R)-3,4-Dihydroxy-6-methyl-5-(((1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl)amino)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glucobay
459080010
459080050
ACRO459080010EA
116
EUR
ACRO459080010
ACRO459080050
Acarbose 95%
Acarbose
Formula:
C₂₅H₄₃NO₁₈ MW: 645,61 g/mol Melting Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00869592 CAS Number: 56180-94-0 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤5 % |
HPLC | ≥94 % |
Specific optical rotation | +168° to +183° (20°C, 589 nm) (1 % in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...