Chelidamic acid hydrate 97%

Supplier: Thermo Scientific
Warning

Synonyms: 1,4-Dihydro-4-oxo-2,6-pyridinedicarboxylic acid hydrate, 4-Oxo-1,4-dihydropyridine-2,6-dicarboxylic acid hydrate

458870010 458870050 458870250
ACRO458870010EA 47.3 EUR
ACRO458870010 ACRO458870050 ACRO458870250
Chelidamic acid hydrate 97%
Chelidamic acid hydrate
Formula: C₇H₅NO₅·xH₂O
MW: 183,12 g/mol (anhy)
Melting Pt: 265…270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066478
CAS Number: 138-60-3

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Water ≤10 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR