4,4',4''-Tri-9-carbazolyltriphenylamine 99%

Supplier: Thermo Scientific
Warning

Synonyms: Tris(4-carbazol-9-ylphenyl)amine, 4,4',4''-Tris(carbazol-9-yl)triphenylamine, Tris(4-(9H-carbazol-9-yl)phenyl)amine, TCTA

459180010 459180050
ACRO459180010EA 87.7 EUR
ACRO459180010 ACRO459180050
4,4',4''-Tri-9-carbazolyltriphenylamine 99%
4,4',4''-Tri-9-carbazolyltriphenylamine
Formula: C₅₄H₃₆N₄
MW: 740,91 g/mol
Melting Pt: 297…300 °C
Storage Temperature: Ambient
MDL Number: MFCD03093250
CAS Number: 139092-78-7

Order Now

Specification Test Results

Appearance White to off-white Powder or crystalline powder
Infrared spectrum Conforms
NMR (1H-NMR) Consistent with structure
Elemental Analysis 7.45 to 7.68 % Nitrogen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR