2-Amino-6-(trifluoromethoxy)benzothiazole 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole
459360010
459360050
459360100
ACRO459360010EA
76.2
EUR
ACRO459360010
ACRO459360050
ACRO459360100
2-Amino-6-(trifluoromethoxy)benzothiazole 98%
2-Amino-6-(trifluoromethoxy)benzothiazole
Formula:
C₈H₅F₃N₂OS Melting Pt: 114…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00210213 CAS Number: 1744-22-5 UN: 2811 ADR: 6.1,II |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 116 to 120 °C |
Water | ≤0.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...