Silver sulphadiazine 98%

Supplier: Thermo Fisher Scientific
Warning

459650050 459650250
ACRO459650050EA 80.8 EUR
ACRO459650050 ACRO459650250
Silver sulphadiazine 98%
Silver sulphadiazine
Formula: C₁₀H₉AgN₄O₂S
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00072101
CAS Number: 22199-08-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR