2-(Hydroxymethyl)indole 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(Indole-2-yl)methanol, 2-Indolmethanol, Indole-2-methanol, Indole-2-carbinol, 1H-Indole-2-methanol, (1H-Indol-2-yl)methanol, 2-Indolcarbinol
459810010
459810050
ACRO459810010EA
63.4
EUR
ACRO459810010
ACRO459810050
2-(Hydroxymethyl)indole 96%
2-(Hydroxymethyl)indole
Formula:
C₉H₉NO MW: 147,18 g/mol Melting Pt: 72…78 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00030195 CAS Number: 24621-70-3 |
Specification Test Results
Appearance | White to light yellow to tan Solid |
Infrared spectrum | Conforms |
Melting point | 72 to 78 °C |
Water | ≤1 % |
GC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...